missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III, IV) oxide, 99% (REO)

Product Code. 11425380
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11425380

Brand: Thermo Scientific Alfa Aesar 044594.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol

Used in ceramic structures. Have a wide range of applications from producing clay bowls to advanced electronics. Praseodymium oxide is also useful in light weight structural components in aerospace and electrochemical applications such as fuel cells due to its ionic conductivity. Component of didymium glass which is a colorant for welder's goggles, also as important additive of Praseodymium yellow pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-29-5
Molecular Formula O11Pr6
Molecular Weight (g/mol) 1021.44
MDL Number MFCD00011178
InChI Key HPZIIFVSYNLWNX-UHFFFAOYSA-N
IUPAC Name hexapraseodymium(3+) undecaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]

Specifications

Melting Point 2500°C
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water and only soluble in strong acid.
Formula Weight 1021.43
Percent Purity 99%
Assay (REO)
Chemical Name or Material Praseodymium (III, IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.