missing translation for 'onlineSavingsMsg'
Learn More
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11396288

Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11396288
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11396288

Brand: Thermo Scientific Alfa Aesar 011235.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-29-5
Molecular Formula O11Pr6
Molecular Weight (g/mol) 1021.44
MDL Number MFCD00011178
InChI Key HPZIIFVSYNLWNX-UHFFFAOYSA-N
IUPAC Name hexapraseodymium(3+) undecaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]

Specifications

Melting Point 2500°C
Color Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water. Moderately soluble in strong mineral acids.
Formula Weight 1021.43
Appearance Black powder
Chemical Name or Material Praseodymium(III, IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT29I022 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog Number011235.22,011235.14
Lot NumberM14I024 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011235.22,011235.14
Lot NumberA16Y024 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011235.22,011235.14
Lot Number23308 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011235.22,011235.14
Lot NumberW27H012 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011235.22,011235.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.