missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III, IV) oxide, REacton™, 99.996% (REO)

Product Code. p-7048568
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11341438

Brand: Thermo Scientific Alfa Aesar 010914.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol

It is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-29-5
Molecular Formula O11Pr6
Molecular Weight (g/mol) 1021.44
MDL Number MFCD00011178
InChI Key HPZIIFVSYNLWNX-UHFFFAOYSA-N
IUPAC Name hexapraseodymium(3+) undecaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]

Specifications

Melting Point 2500°C
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water; moderately soluble in strong mineral acids.
Formula Weight 1021.43
Percent Purity 99.996%
Grade Reacton
Assay (REO)
Chemical Name or Material Praseodymium(III, IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.