missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO)

Product Code. p-7048687
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11346298

Brand: Thermo Scientific Alfa Aesar 011240.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15878-77-0 | H12N3O15Pr | 435.01 g/mol

Praseodymium(III) nitrate hexahydrate is used in electrical components, optical glasses and photo-optical materials. Also find use as catalysts and laboratory reagent. In addition, it is used in the solvothermal process for formation of doped lanthanide oxysulfide. It is a precursor for the electrodeposition of praseodymium oxide for electrochemical applications. It is a starting material for the preparation of ceramic pigments, metal-organic frameworks and mixed conductors. It finds utility in the synthesis of other praseodymium compounds and complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00149826
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
IUPAC Name praseodymium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Solubility Information Soluble in water and alcohol.
Formula Weight 435.01 (326.92 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Praseodymium(III) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Praseodymium(III) nitrate hexahydrate, REacton
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Praseodymium(III) nitrate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.