missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) nitrate pentahydrate, 99.9%, (trace metal basis)

Product Code. 10522601 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 10522601

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14483-17-1 | N3O9Pr·5H2O | 417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 14483-17-1
Color Green
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.90%
Molecular Formula N3O9Pr·5H2O
Linear Formula Pr(NO3)3·5H2O
MDL Number MFCD00149825
Molecular Weight (g/mol) 417
Formula Weight 417
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Praseodymium(III) nitrate pentahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Praseodymium(III)-nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.