Learn More
Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
£102.51 - £283.00
Chemical Identifiers
CAS | 52093-27-3 |
---|---|
Molecular Formula | C3F9O9PrS3 |
Molecular Weight (g/mol) | 588.097 |
MDL Number | MFCD00192417 |
InChI Key | ROUBZIWQWFQCHU-UHFFFAOYSA-K |
Synonym | praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate |
PubChem CID | 9894708 |
IUPAC Name | praseodymium(3+);trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313868
|
Thermo Scientific Alfa Aesar
040310.09 |
10 g |
£102.51
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323868
|
Thermo Scientific Alfa Aesar
040310.18 |
50 g | N/A | ||||||
Description
It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52093-27-3 | |
588.097 | |
ROUBZIWQWFQCHU-UHFFFAOYSA-K | |
9894708 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3] |
C3F9O9PrS3 | |
MFCD00192417 | |
praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate | |
praseodymium(3+);trifluoromethanesulfonate |
Specifications
52093-27-3 | |
C3F9O9PrS3 | |
praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate | |
ROUBZIWQWFQCHU-UHFFFAOYSA-K | |
praseodymium(3+);trifluoromethanesulfonate | |
9894708 | |
98% |
Powder | |
MFCD00192417 | |
Insoluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3] | |
588.097 | |
588.11 | |
Praseodymium(III) trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only