missing translation for 'onlineSavingsMsg'
Learn More
Praseodymium ingot, 99% (REO), Thermo Scientific Chemicals Product Code.: 11336507

Praseodymium ingot, 99% (REO), Thermo Scientific Chemicals

Product Code. 11336507
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11336507

Brand: Thermo Scientific Alfa Aesar 000267.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Gray to Silver
Appearance Gray to silver ingot
Assay Percent Range 99.9%
Quantity 10 g
Odor Odorless
Sensitivity Air and moisture sensitive
Chemical Name or Material Praseodymium, ingot
compliance-icons
Product Identifier
  • Praseodymium ingot
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium ingot, 99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.