missing translation for 'onlineSavingsMsg'
Learn More
Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 11303848

Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11303848
50 mm
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy

Product Code. 11303848

Brand: Thermo Scientific Alfa Aesar 040296.DJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Quantity 50 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.86g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Praseodymium rod, 12.7mm (0.5 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX26H001 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040296.HF,040296.DH,040296.DJ,040296.DM
Lot NumberX16D064 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number040296.HF,040296.DH,040296.DJ,040296.DM
Lot NumberN25H008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040296.HF,040296.DH,040296.DJ,040296.DM
Lot NumberG17K38 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040296.HF,040296.DH,040296.DJ,040296.DM
Lot NumberG15M21 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040296.HF,040296.DH,040296.DJ,040296.DM

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.