missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta), Thermo Scientific Chemicals
Form: Approx. 0.22g/mm, Cast rod, diameter is approximate
£107.54 - £544.71
Chemical Identifiers
CAS | 7440-10-0 |
---|---|
Molecular Formula | Pr |
Molecular Weight (g/mol) | 140.91 |
MDL Number | MFCD00011174 |
InChI Key | PUDIUYLPXJFUGB-UHFFFAOYSA-N |
Synonym | unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg |
PubChem CID | 23942 |
ChEBI | CHEBI:49828 |
IUPAC Name | praseodymium |
SMILES | [Pr] |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
11333838
|
Thermo Scientific Alfa Aesar
040295.HF |
10 mm |
£107.54
10mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363838
|
Thermo Scientific Alfa Aesar
040295.DH |
25 mm |
£171.86
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353838
|
Thermo Scientific Alfa Aesar
040295.DJ |
50 mm |
£296.48
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343838
|
Thermo Scientific Alfa Aesar
040295.DM |
100 mm |
£544.71
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Especificaciones
7440-10-0 | |
Cast Rod | |
Pr | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
Silver colored rod |
Silver | |
99.10% | |
MFCD00011174 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
Odorless | |
Praseodymium |
Seguridad y manipulación
missing translation for 'einecsNumber' : 231-120-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only