missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pravastatin sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Water-soluble, competitive inhibitor of 3-hydroxy-3-methyl coenzyme A (HMG-CoA) reductase. Potently blocks cholesterol synthesis in vivo (Ki∽ 1 nM) and displays cardioprotective properties.
Specifications
Specifications
CAS | 81131-70-6 |
Molecular Formula | C23H35NaO7 |
MDL Number | MFCD00887601,MFCD00887601 |
Quantity | 50 mg |
Synonym | pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol |
InChI Key | VWBQYTRBTXKKOG-IYNICTALSA-M |
SMILES | [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 |
IUPAC Name | sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
Molecular Weight (g/mol) | 446.52 |
PubChem CID | 16759173 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction