missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PRDM4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16852184
|
Proteintech
13792-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16842184
|
Proteintech
13792-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.Specifications
PRDM4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q80V63, Q9QZP2, Q9UKN5 | |
11108, 170820, 72843 | |
PRDM4 Fusion Protein Ag4580 | |
Primary | |
-20°C | |
PRDM4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PFM1, PR domain containing 4, PR domain containing protein 4, PRDM4 | |
Prdm4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |