missing translation for 'onlineSavingsMsg'
Learn More

PRE-084 hydrochloride, Tocris Bioscience™

Product Code. 30001102
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective σ1 agonist

High affinity, selective σ1 agonist (Ki values are 2.2 and 13091 nM for σ1 and σ2 receptors respectively). Selective over PCP receptors (IC50 > 100000 nM) and several other receptor systems. Produces antiamnesic and antitussive effects in vivo. Also exhibits protective effects in PC6.3 cells expressing mutant huntingtin by activating NF-κB signaling.

Chemical Identifiers

CAS 138847-85-5
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.43
MDL Number MFCD00673883
InChI Key RQHKZUBCUZVZEF-UHFFFAOYSA-N
Synonym pre-084 hydrochloride, 2-morpholin-4-yl ethyl 1-phenylcyclohexane-1-carboxylate hydrochloride, pre-084, solid, 2-4-morpholinethyl 1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinethyl-1-phenylcyclohexanecarboxylate hydrochloride, 2-4-morpholinyl ethyl 1-phenylcyclohexanecarboxylate hydrochloride
PubChem CID 11314197
IUPAC Name 2-(morpholin-4-yl)ethyl 1-phenylcyclohexane-1-carboxylate
SMILES O=C(OCCN1CCOCC1)C1(CCCCC1)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 353.89
Percent Purity >99%
Chemical Name or Material PRE-084 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.