Learn More
Fisherbrand™ Pre-Filled Bead Mill Tubes
For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.
£138.00 - £222.00
Product Code | Brand | Capacity (Metric) | Particle Size | Material | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Particle Size | Material | Product Type | Price | Quantity & Availability | ||
15545799
|
Fisherbrand™
|
2 mL | 0.5mm | Glass | Tough Microorganism Lysing Mix Tubes |
£175.00
Each |
In Stock
Log in to see stock. |
||
15555799
|
Fisherbrand™
|
2 mL | 1.4 mm | Ceramic | Soft Tissue Homogenizing Mix Tubes |
£175.00
Pack of 50 |
In Stock
Log in to see stock. |
||
15535799
|
Fisherbrand™
|
2 mL | 2.4 mm | Metal | Hard Tissue Grinding Mix Tubes |
£175.00
Each |
In Stock
Log in to see stock. |
||
15565799
|
Fisherbrand™
|
2 mL | 2.8 mm | Ceramic | Hard Tissue Homogenizing Mix Tubes |
£175.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
15585799
|
Fisherbrand™
|
7 mL | 1.4 mm | Ceramic | Soft Tissue Homogenizing Mix Tubes |
£203.00
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||
15575799
|
Fisherbrand™
|
7 mL | 2.4 mm | Metal | Hard Tissue Grinding Mix Tubes |
£138.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||
15595799
|
Fisherbrand™
|
7 mL | 2.8 mm | Ceramic | Hard Tissue Grinding Mix Tubes |
£222.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
Description
- Polypropylene tubes closed with screw caps
- Non-Sterile