Learn More
3M™ Pre-filter Pads
Extends the life of your Powered Air Respirator by capturing large particles that can clog a filter.
Brand: 3M™ 7000006600
18.19 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : S8
Additional Details : Weight : 0.30600kg
Description
- Frequent changing of prefilters can optimize performance
- For use with 3M Jupiter™ Powered Air Respirators
Specifications
Prefilter Pad | |
Jupiter Powered Air Turbo Unit | |
461-00-02P24X5 | |
24/Pk. |