missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Pre-miR™ miRNA Precursor

Product Code. 10782307
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10782307

Brand: Ambion™ AM17100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Pre-miR™ miRNA Precursor

miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
Description Pre-miR™ miRNA Precursor Molecule, 5nmol, Liquid, Protect Against Physical Damage, Isolate From Incompatible Substances, Store in Original Container
Final Product Type Pre-miR™ miRNA Precursor Molecule
Packaging Type 1 Each
Product Line Ambion, Pre-miR
Product Type Pre-miR™ miRNA Precursor Molecule
Quantity 5 nmol
Sample Size 5nmol
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.