missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pre-miR™ miRNA Precursor Negative Control #1

Product Code. 10249444
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10249444

Brand: Invitrogen™ AM17110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Pre-miR™ miRNA Precursor Negative Control #1

Pre-miR™ miRNA Precursors are designed to mimic endogenous miRNAs. Pre-miR miRNA Precursor Negative Control #1 is designed to use as a negative control for experiments using Pre-miR miRNA Precursors. Transfect Pre-miR miRNA Precursor Negative Control #1 using the same methodology as for your experimental Pre-miR miRNA Precursors, then use the target gene expression from the sample transfected with the Pre-miR miRNA Precursor Negative Control #1 as a baseline for evaluation of the effect of the experimental Pre-miR miRNA Precursor on target gene expression. Advantages of the Pre-miR™ miRNA Precursor Negative Control #1:

  • Optimized design results in correct strand utilization by RISC
  • Purified and ready to transfect

    About Pre-miR miRNA Precursors
    Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments.

    Using Pre-miR miRNA Precursor Negative Control #1
    Pre-miR™ Negative Control #1 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. RNA oligonucleotides are susceptible to degradation by exogenous ribonucleases introduced during handling. Wear gloves when handling this product. Use RNase-free reagents, tubes, and barrier pipette tips. Upon receipt, store in a non-frost-free freezer at or below -20°C (dried oligonucleotides are shipped at ambient temperature).

    NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
    Format Tube
    Control Type Negative Control
    RNAi Type miRNA Mimic
    For Use With (Application) miRNA & Non-Coding RNA Analysis
    Label or Dye Unconjugated
    Product Line Ambion, Pre-miR
    Product Type miRNA Mimic
    Quantity 5 nmol
    Shipping Condition Room Temperature

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.