Learn More
LTE Scientific™ Pre/Post Vacuum system booster heaters
For use with Touchclave-R. LTE Scientific™ Pre/Post Vacuum system booster heaters are for use with Touchclave-R suitable for liquids, no vacuum air removal, and no autofill.
Brand: LTE Scientific™ TCR/120/V5
Code : NEW
Additional Details : Weight : 265.00000kg
Description
- For use with Touchclave-R
Specifications
Touchclave-R Autoclaves |