missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prealbumin/transthyretin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11891-1-AP-20UL
This item is not returnable.
View return policy
Description
Transthyretin, one of the three prealbumins including alpha-1-antitrypsin, transthyretin and orosomucoid. Transthyretin is a carrier protein; it transports thyroid hormones in the plasma and cerebrospinal fluid, and also transports retinol (vitamin A) in the plasma. The protein consists of a tetramer of identical subunits. More than 80 different mutations in this gene have been reported; most mutations are related to amyloid deposition, affecting predomitly peripheral nerve and/or the heart, and a small portion of the gene mutations is non-amyloidogenic. The diseases caused by mutations include amyloidotic polyneuropathy, euthyroid hyperthyroxinaemia, amyloidotic vitreous opacities, cardiomyopathy, oculoleptomeningeal amyloidosis, meningocerebrovascular amyloidosis, carpal tunnel syndrome, etc.Specifications
Prealbumin/transthyretin | |
Polyclonal | |
Unconjugated | |
TTR | |
ATTR, HsT2651, PALB, Prealbumin, TBPA, transthyretin, TTR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7276 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02766 | |
TTR | |
Prealbumin/transthyretin Fusion Protein Ag2499 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |