missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREB Mouse anti-Human, Unconjugated, Clone: 2F6A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PREB |
---|---|
Clone | 2F6A1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849325
|
Proteintech
66981-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839325
|
Proteintech
66981-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PREB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PREB. This antibody recognizes human antigen. The PREB antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PREB | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
10113 | |
Primary |
2F6A1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG1 |