Learn More
Bochem™ Precision Forceps
Product Code. 11895151
Click to view available options
Length (Metric):
120 mm
Unit Size:
Pack of 10
Product Code. 11895151
Brand: Bochem™ 1640
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel
  • Points without ridges
  • Self-tensing
Specifications
Tip Style Sharp
No. per Pack 1 Pack
Material Stainless Steel
Length (Metric) 120 mm
Product Type Forceps, Precision
Type Forceps