Learn More
Thermo Scientific™ Precision™ Shaking Water Baths
Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.
£1903.72 - £3097.75
Specifications
Chamber Material | 304 Stainless Steel |
---|---|
Controller Type | Digital |
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Product Code | Brand | Model | Description | Capacity (Metric) | Electrical Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Description | Capacity (Metric) | Electrical Requirements | Price | Quantity & Availability | ||
15365867
|
Thermo Scientific™
TSSWB15 |
Precision SWB 15 | 15L Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz |
£1960.00
Each |
In Stock
Log in to see stock. |
||
15375867
|
Thermo Scientific™
TSSWB27 |
Precision SWB 27 | 27L Shaking Water Bath | 27 L | Global Voltage 100/115 V 200/230 V 50/60 Hz |
£3097.75
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
15385867
|
Thermo Scientific™
TSSWB15S |
Precision SWB 15S | 15L Shallow Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz |
£1903.72
Each |
In Stock
Log in to see stock. |
||
15395867
|
Thermo Scientific™
TSDUB15 |
Precision DUB 15 | 15L Dubnoff Shaking Water Bath | 15 L | Global Voltage 100/115 V 200/230 V 50/60 Hz |
£2576.17
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
Description
Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere
- Easily clean and maintain bath with coil-free design
- Optimize scheduling with auto-on and auto-off timers
- Help prevent bath overheating and damage with low-fluid protection
- Conveniently save commonly used settings with four temperature and shaking speed presets
- Help protect your work with audible alarms
- Easily operate and monitor with icon-based graphical display
- Accommodate taller labware with new hinged lid and extended height
- Features next-generation shaker motor
- Warranty varies by country, please contact us for details
Specifications
304 Stainless Steel | |
UL, CE | |
Monochrome LCD | |
4 | |
±0.05°C |
Digital | |
Ambient +5° to 100°C | |
Epoxy Powder-coated Cold Rolled Steel | |
±0.1°C | |
Global Voltage 100/115 V 200/230 V 50/60 Hz |