missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Precision Tex 2 Gloves
Click to view available options
10
Description
- Mixed grain leather
- Green cotton interlock back
- Elastic tightening
- Gunn cut
- Winged thumb
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance (Europe) | EN 420-2003 Basic Standards, EN 388-2016 Mechanical Hazards |
Classification | Dexterity: 5 |
Color | White, Green |
Hand | Specific |
Material | Mixed Grain Leather, Cotton |
Product Type | Leather Palm Gloves |
Size | 10 |
Style | Gunn |
For Use With (Application) | Automotive, Aviation, Iron, Steel, Logistics, Maintenance, and Transportation |
Glove Cuff Style | Open |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'