missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Predictor™ hERG Fluorescence Polarization Assay Kit

Product Code. 10532304
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10532304

Brand: Invitrogen™ PV5365

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy
For Research Use Only. All usage must comply with product instructions.
 

Predictor™ hERG Fluorescence Polarization Assay Kit

The Predictor™ hERG Fluorescence Polarization Assay Kit provides a set of validated components to perform hERG channel biochemical binding studies in the absence of radioligand. The assay was designed to identify potential hERG channel blockers by producing data that accurately correlates with patch-clamp electrophysiology studies. The assay is based on the principle of fluorescence polarization where a red-shifted fluorescent tracer is displaced from the hERG channel by compounds that bind to the channel. Assay performance was validated with a panel of established hERG channel blockers. Results from the Predictor™ assay demonstrate a high correlation with those obtained from patch clamp techniques.
TRUSTED_SUSTAINABILITY

Specifikationer

Color Red
Conjugate Predictor hERG Tracer Red
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Quantity 1 Kit
Packaging 384-well Plate
Content And Storage

PV5365 Predictor™ hERG Fluorescence Polarization Assay Kit
(Other materials are required separately; please refer to the protocol)
Includes:
• Predictor™ hERG Membranes
• Predictor™ hERG Tracer Red
• Predictor™ hERG Buffers
• E-4031
• Predictor™ hERG Fluorescence Polarization Assay Protocol
• Certificate of Analysis
Store the Predictor™ hERG Membranes at -80°C. The Predictor™ Tracer Red, Predictor™buffers, and E-4301 should be stored at -20°C.

Assay Entry Biochemical binding
Druggable Target Ion Channels
Gene Symbol hERG
Readout End Point
Shipping Condition Dry Ice
System Type Predictor™ System
Target Entry hERG
Product Line Predictor
Protein Family Ion Channels
Product Type Assay Kit
Visa mer Visa mindre

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.