Learn More
Prednisolone, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Prednisolone is a derivative of prednisone with an oxo to beta-hydroxy reduction at position 11
• Prednisolone binds cell surface receptors and is transported to the nucleus, where it binds certain nuclear receptors responsible for transcription of proinflammatory cytokines
Applications
• Prednisolone can be used for laboratory research on tumor growth and prevention
• Prednisolone may also be used for developmental and endocrine research, in vivo or in vitro
Specifications
Specifications
Melting Point | 240.0°C |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Merck Index | 15,7841 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in methanol, dioxane, slightly soluble in acetone, chloroform,, ethylacetate |
Formula Weight | 360.44 |
Percent Purity | 99% |
Chemical Name or Material | Prednisolone |
Safety and Handling
- Prednisolone
- Danger
- Reproductive toxicity Category 1B
- H360D-May damage the unborn child.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only