Learn More
Prednisone, 98%, Thermo Scientific Chemicals
Prednisone, 98%, CAS # 53-03-2, is a synthetic glucocorticoid with anti-inflammatory, immunomodulatory, antineoplastic, and adrenergic activities.
£82.80 - £284.00
Chemical Identifiers
CAS | 53-03-2 |
---|---|
Molecular Formula | C21H26O5 |
Molecular Weight (g/mol) | 358.434 |
InChI Key | XOFYZVNMUHMLCC-ZPOLXVRWSA-N |
Synonym | prednisone, deltasone, dehydrocortisone, decortin, meticorten, orasone, prednison, rectodelt, ultracorten, dacortin |
PubChem CID | 5865 |
ChEBI | CHEBI:8382 |
IUPAC Name | (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione |
SMILES | CC12CC(=O)C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13458638
|
Thermo Scientific Acros
448950050 |
5 g | Glass bottle |
£82.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13468638
|
Thermo Scientific Acros
448950250 |
25 g | Glass Bottle |
£284.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Prednisone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities
• It shows antineoplastic and adrenergic activities against hormone-sensitive cancer cells
• This compound binds to its receptors and enters the nucleus activating specific nuclear receptors, resulting in an altered gene expression and inhibiting proinflammatory cytokine production
Applications
• Prednisone can reduce circulating lymphocytes, promote cell differentiation, and stimulate apoptosis in sensitive tumor cell lines
• It is considered a prodrug that is converted by the liver into prednisolone which is the active steroid compound
• It is also used in some studies to investigate the effects of daily administration of MIDD0301, on the animal and organs weights and hematological profile
Chemical Identifiers
53-03-2 | |
358.434 | |
prednisone, deltasone, dehydrocortisone, decortin, meticorten, orasone, prednison, rectodelt, ultracorten, dacortin | |
CHEBI:8382 | |
CC12CC(=O)C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C |
C21H26O5 | |
XOFYZVNMUHMLCC-ZPOLXVRWSA-N | |
5865 | |
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione |
Specifications
53-03-2 | |
C21H26O5 | |
XOFYZVNMUHMLCC-ZPOLXVRWSA-N | |
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione | |
5865 | |
358.43 |
98% | |
prednisone, deltasone, dehydrocortisone, decortin, meticorten, orasone, prednison, rectodelt, ultracorten, dacortin | |
CC12CC(=O)C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C | |
358.434 | |
CHEBI:8382 | |
Prednisone |
Safety and Handling
- Prednisone
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only