missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex Preformed Mask silv-Air c 2220 FFP2
The silv-Air c product family consists of classic folding and cup-shaped face masks which fit different face shapes. Headband lacing system and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
Brand: Uvex 8732220
This item is not returnable.
View return policy
Description
General Features
- Particle-filtering preformed mask with exhalation valve
- Color: silver
Protection Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
- Additional activated carbon filter against odors, gases and vapors below threshold
Comfort features
- Seamless headband and flexible, adjustable nose clip for a comfortable, secure fit
- Comfortable sealing lip in nose area for secure fit
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
Specifications
EN 149 (D marked) | |
Single-use | |
Standard | |
FFP2 |
Cup/Molded | |
Polypropylene, Polyester | |
Yes | |
Non-sterile |