missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregnenolone, 99%, Thermo Scientific Chemicals
£38.70 - £371.00
Chemical Identifiers
CAS | 145-13-1 |
---|---|
Molecular Formula | C21H32O2 |
Molecular Weight (g/mol) | 316.47 |
MDL Number | MFCD00003628 |
InChI Key | ORNBQBCIOKFOEO-OYZZJKLHSA-N |
Synonym | 3-hydroxypregn-5-en-20-one |
PubChem CID | 44307923 |
IUPAC Name | 1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone |
SMILES | CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582951
|
Thermo Scientific Acros
164420050 |
5 g | Glass bottle |
£38.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10583141
|
Thermo Scientific Acros
164420250 |
25 g | Glass bottle |
£130.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10443965
|
Thermo Scientific Acros
164421000 |
100 g | Glass bottle |
£371.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
145-13-1 | |
316.47 | |
ORNBQBCIOKFOEO-OYZZJKLHSA-N | |
44307923 | |
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C |
C21H32O2 | |
MFCD00003628 | |
3-hydroxypregn-5-en-20-one | |
1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone |
Specifications
145-13-1 | |
White | |
Authentic | |
C21H32O2 | |
15, 7851 | |
+ 28.5 | |
Solubility in water: slightly soluble. Other solubilities: solubility (g/100 mL of solution):,carbon terachloride 0.5 / petroleum ether 0.1 / et,acetone 0.6 / chloroform 17.0 / ethanol 1.9 / benz,isopropanol 1.5,solubility (g/100 mL of solvent): | |
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C | |
316.47 | |
316.47 | |
Powder |
190°C to 193°C | |
0.5% max. | |
98.5% min. (GC) | |
MFCD00003628 | |
+28.50° (20°C c=1,EtOH) | |
3-hydroxypregn-5-en-20-one | |
ORNBQBCIOKFOEO-OYZZJKLHSA-N | |
1-[(10R,13S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone | |
44307923 | |
99% | |
Pregnenolone |
Safety and Handling
missing translation for 'einecsNumber' : 205-647-4
RUO – Research Use Only