Learn More
Pacific Handy Cutter Premium Multi Functional Safety Knife with Guard & Springback Blade
The S7 Safety Cutter combines the utility of an industrial knife with the safety benefits of a self-retracting safety cutter. To maximize productivity this 3-in-1 tool incorporates an easy release film cutter, heavy duty tape splitter and versatile safety box cutter.
Brand: Pacific Handy Cutter S7
Additional Details : Weight : 0.00680kg
Description
- To prevent painful and costly injuries, the ambidextrous S7 features an innovative robust metal guard that moves in tandem with the blade, retracting both the guard and blade simultaneously.
- The guard covers the blade during top cutting to protect the user and the unique angle of the blade is set specifically to avoid damaging the contents within the box.
- The spring-back blade can also be extended without the guard when needed and has three locking blade depth positions; top cut, shallow tray cut and deep cut.
- The integrated film cutter is perfect for safely cutting shrink-wrap, plastic straps, twine and more.
- Users also find that utilizing the heavy duty tape splitter while opening boxes further prevents injuries and extends the life of the blades.
- With the easiest blade change on the market this universal cutting tool reduces costly razor blade injuries and makes box cutting safe, easy and comfortable.
- For better retention, purchase with corresponding UKH-545 Safety Holster and CL-36 Coil Lanyard.
Specifications
Safety Knife | |
Non-sterile | |
Blue |
No | |
Guard and Springback Blade |