missing translation for 'onlineSavingsMsg'
Learn More

Kappes Systeme™ Premium Storage Bins

Brand:  Kappes Systeme™ 6002.00.0250

Additional Details : Weight : 0.59400kg

 View more versions of this product

Product Code. 12672055

  • POA / Each
Estimated Shipment: 05-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »


Description

Description

Specifications

Specifications

Storage Bin
Blue
Size 3
Food Storage
200 mm
Polyethylene
Rectangular
Feed and Roller Conveyor
350 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers