missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Premo™ Halide Sensor

Premo™ Halide Sensor

Brand:  Molecular Probes™ P10229

647.52 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10597015

  • £852.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The Premo™ Halide Sensor is based on a yellow fluorescent protein (YFP) molecule sensitive to halide ions. The combination of the YFP (Venus) biosensor with efficient and non-cytopathic BacMam delivery and expression gives researchers a highly sensitive, robust and easy-to-use tool to efficiently screen halide ion channels and transporter modulators in their cellular models of choice. No specialized instruments are needed, as the sensor works on standard HTS platforms. - Cl– channel–specific—measure halide ion flux in voltage- and ligand-gated chloride channels and transporters - Fast—measure chloride flux in high-throughput mode with highly reproducible results - Robust—reliably high expression of the bright and fast maturating Venus sensor gives an excellent signal window - Convenient—study chloride flux in your cellular model by efficient and non-cytopathic delivery of Premo™ Halide Sensor by BacMam technology - Pharmacologically relevant—known modulators show dose-dependent quenching and BacMam delivery enables assays in primary cells The assay combines the YFP Venus halide sensor with a surrogate ion for chloride (iodide); upon stimulation of the chloride channel or transporter, iodide ions flow down the concentration gradient into the cells and quench YFP fluorescence upon binding; the amount of quench is directly proportional to the ion flux (chloride channel or transporter activity). Due to the bright Venus fluorescence, the Premo™ Halide Sensor signal window is excellent. We have demonstrated the utility and efficiency of the Premo™ Halide Sensor as a robust and true “no wash” HTS chloride assay. Premo™ Halide Sensor is pre-packaged and ready for immediate use. It contains all components required for cellular delivery and expression. Screening can be conducted in complete medium and without any wash steps. To uncouple cell maintenance and preparation from screening, simply freeze transduced cells and bring out as little as 4 h before the screen. Both stable cell lines and human primary cells can be prepared frozen and “assay-ready”.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yellow
Baculovirus (BacMam System)
Cell Viability and Proliferation
Halide Sensor
Visible
Molecular Probes™, Premo™
Wet Ice
The Premo™ Halide Sensor is a pharmacologically relevant sensor for functional studies of ligand- and voltage-gated chloride channels. It is based on a halide-sensitive form of Venus, the brightest available YFP molecule, affording an excellent signal window. Packaged in BacMam format, delivery is efficient in a range of cell types, including human primary cells. We have demonstrated the utility and efficiency of the Premo™ Halide Sensor as a robust and true “no wash” HTS chloride assay.
Premo™ Halide Sensor should be stored at 4°C, protected from light and is supplied with an enhancer (store at 4°C). When stored as directed, the product is stable for 6 months.
Fluorescence
Fluorescence Microscope
Protein-Based
Liquid
20 mL kit
Ion Channels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2983225 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberP10229
Lot Number2896530 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog NumberP10229
Lot Number2806833 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog NumberP10229
Lot Number2697983 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog NumberP10229
Lot Number2626236 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog NumberP10229
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Premo™ Halide Sensor > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.