missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepMan™ Ultra Sample Preparation Reagent

Product Code. 30000667 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30000667

Brand: Applied Biosystems™ 4318930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PrepMan™ Ultra Sample Preparation Reagent was developed for the rapid preparation of DNA template from Gram-negative food-borne pathogens for use in PCR amplification reactions.

The PrepMan™ Ultra Sample Preparation Reagent was developed for the rapid preparation of DNA template from Gram-negative food-borne pathogens for use in PCR amplification reactions. These samples often have high lipid content that can inhibit PCR amplification of the template. Using a simple boil and spin protocol, PrepMan™Ultra Reagent efficiently inactivates PCR inhibitors, significantly reducing the need to repeat the template preparation step.

PrepMan™ Ultra reagent has been used to prepare PCR-ready template from a wide variety of different sources, including bacteria, yeast, filamentous fungi, both from a plate or from liquid culture, mammalian tissue smears, human cells (buccal swab), and mammalian whole blood. The resulting template has been used amplified in allelic discrimination assays, genetically modified organism (GMO) detection and quantification, food pathogen detection, microbial identification, and bacterial strain typing.

• Simple, rapid protocol reduces time to results.
• Unique formulation inactivates PCR inhibitors, yielding consistent results.
• PCR-ready template can be prepared from a wide range of sample types.
• Affordable method with a low price per preparation.
• Homogeneous solution with minimal sample-handling requirements.
• Microcentrifuge tube-based format means no additional equipment is needed for template preparation.

PrepMan™ Ultra reagent is a novel formulation and is a homogeneous solution that does not contain Chelex™ or any other type of resin or matrix.

This product is a replacement⁄improvement of PrepMan™ Sample Preparation Reagent, and should be recommended to current and potential PrepMan™ Reagent users.

Used With
3500/3500xL Genetic Analyzer Capillary Array (50 cm), 3130xL/3100 Genetic Analyzer Capillary Array (36 cm), 96-Well Plate Septa, AmpliTaq Gold™ PCR Master Mix, BigDye™ Terminator v1.1 Cycle Sequencing Kit, POP-6™ Polymer for 3100/3100-Avant™ Genetic Analyzers, PrepMan™ Ultra Sample Preparation Reagent, TaqMan™ GMO Maize 35S Detection Kit, TaqMan™ GMO Soy 35S Detection Kit.

Specifications

Content And Storage Contents: One bottle of reagent.

Store the PrepMan Ultra Sample Preparation Reagent, at room temperature. Do not freeze or autoclave the PrepMan Ultra Sample Preparation Reagent.
Format Bottle
Sample Type Bacterial Culture, Food, Fungi, Whole Blood
Final Product Type Genomic DNA, Bacterial DNA
For Use With (Application) Real-Time Quantitative PCR (qPCR), Sequencing, PCR
No. of Reactions 200 Preps
Product Line PrepMan
Quantity 1 kit
Shipping Condition Room Temperature
System Type MicroSEQ™ ID
Test Time 15 min
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.