missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Express Nucleic Acid Extraction Kit

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.

Brand:  Applied Biosystems™ 4466351

Product Code. 30000678

  • £988.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.

Specifications

Specifications

Contains pre-filled reagent cartridges and plastics to perform extraction of DNA and RNA.

Store at room temperature.
Biopharmaceutical Purification Sample, Bulk Drug Substance Sample, Cell Cultures
7500 Fast System, AutoMate Express™ Nucleic Acid Extraction System
Sufficient for isolation of 52 samples
PrepSEQ™
Room Temperature
≤ 1 hr.
Magnetic Bead
Genomic DNA, Total RNA
Automated Protocols
Real-Time Quantitative PCR (qPCR)
1 Kit
Genomic DNA, Total RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.