missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ PrepSEQ™ Lysis Buffer
Shop All Applied Biosystems Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
This PrepSEQ Lysis Buffer is used in the large-scale protocol of the PrepSEQ Express Nucleic Acid Extraction Kit (Cat. No. 4466351). It is not necessary for the standard protocol (sufficient lysis buffer is included in the kit's lysis tubes). As with other products in the Pharma Analytics Sample Preparation product line, PrepSEQ Lysis Buffer:
- Is completely supported by our global manufacturing and distribution centers
- Has a Certificate of Analysis available
- Has a shelf-life of 12 months
Specifications
Specifications
| Content And Storage | 1 x 50 mL bottle, store at room temperature. |
| Format | Bottle |
| Product Line | PrepSEQ |
| Quantity | 50 mL |
| Reagent Type | Cell Lysis Buffer |
| Shipping Condition | Room Temperature |
Safety and Handling
Product Identifier
- PrepSEQ Lysis Buffer
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Thiocyanic acid
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction