Learn More
Applied Biosystems™ PrepSEQ™ Nucleic Acid Extraction Kit for Food and Environmental Testing
PrepSEQ™ Nucleic Acid Extraction Kit for Food and Environmental Testing
Brand:  Applied Biosystems™ 4480466
Code : NEW
Additional Details : Weight : 0.00001kg
Product Code. 13483119
  • £700.00 / Each
Estimated Shipment: 23-07-2024
to see stock.
Description
Description
The PrepSEQ™ Nucleic Acid Extraction Kit produces high-quality bacterial DNA samples for PCR-based detection from a wide range of food and environmental samples. The quantity and quality of the DNA can greatly impact the success of the sample analysis process and the overall quality of the final result. Food and environmental samples can be challenging to process because they often contain PCR inhibitors. For laboratories looking to achieve better real-time PCR results and improve overall efficiency, the PrepSEQ™ Nucleic Acid Extraction Kit meets these needs.
Automated Sample Preparation for PCR-Based Assays
  • Extract high-quality bacterial DNA
  • Process a wide range of food and environmental samples
  • Fast time-to-results
  • Prepare hundreds of samples per shift per instrument
    High-Throughput Food and Environmental Sample Preparation
    PCR-based bacterial assays have become an important tool in food safety and environmental testing. The Applied Biosystems PrepSEQ™ Nucleic Acid Extraction Kit prepares high-quality bacterial DNA from food and environmental samples for PCR-based testing using magnetic bead-based extraction. In combination with the automated MagMAX™ Express-96 Deep Well Magnetic Particle Processor, this optimized solution enables laboratories to prepare a wide range of sample types at rates of up to hundreds of samples per eight-hour shift.
    Streamlined Workflow
    An easy-to-use, easy-to-learn protocol simplifies integration of the PrepSEQ™ Nucleic Acid Extraction Kit into the laboratory workflow. The same protocol can be used on a wide range of sample types, and the MagMAX™ Express-96 instrument automates many of the workflow steps to reduce hands-on time.
    A Complete Solution
    The PrepSEQ™ Nucleic Acid Extraction Kit is part of a fully integrated, start-to-finish solution for food and environmental testing. The kit has been performance-verified for food and environmental testing with MicroSEQ™ Microbial Detection Kits, the Applied Biosystems™ 7500 Fast Real-Time PCR System, and RapidFinder™ Express Software.
    For research use only. Not intended for any animal or human therapeutic or diagnostic use.
  • Specifications
    Specifications
    Room temperature
    96-well Plate
    Bacteria, Food and Environmental Samples, Liquid Samples (e.g. Serum)
    MagMAX™ express-96 deep well magnetic particle processor
    2 Bottles of lysis buffer, 2 tubes of magnetic particles (store at 2 to 8°C), 1 empty bottle for binding solution, 2 bottles of wash buffer concentrate, 1 bottle of elution buffer, 1 bottle of proteinase K buffer, 1 tube of proteinase K (store at –20°C)
    1 Kit(s)
    1mL
    PrepSEQ™ Nucleic Acid Extraction Kit, 100 preps, Automated Protocols, High Throughput-Compatible, For DNA Extraction, Diagnostic Testing, Bacterial DNA, For PCR, Real-Time Quantitative PCR (qPCR), Food and Environmental Testing
    Enzymatic, Magnetic Bead
    Nucleic Acid Extraction Kit
    For PCR, real-time quantitative PCR (qPCR), food and environmental testing
    100 Preps
    Nucleic Acid Extraction Kit
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    For Testing of Food and Environmental Samples Only.