Learn More
JULABO PRESTO Process Systems
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
Brand: JULABO 9420701.N1
Additional Details : Weight : 150.00000kg
Description
- Wide working temperature ranges without changing fluids
- Ethernet interface
- SD-Card slot
- pace optimized design creates more room directly next to the units
- Connections for USB, Ethernet, RS232, and Alarm output
Specifications
-75°C to +250°C | |
29.3 x 22.4 x 34.6 in. | |
A70 | |
Process System |