Learn More
Julabo™ PRESTO Temperature Control System
Highly precise temperature control and rapid temperature change capacity
Brand:  Julabo™ 9420801.T
Product Code. 15244363
  • £41755.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Julabo PRESTO™ units offer highly precise temperature control and rapid temperature change capacity, making them ideal for applications such as reactor temperature control, substance testing or temperature simulation studies. Highly efficient components permit extremely fast compensation for both exothermic and endothermic reactions. Laboratory users benefit from high flow rates, constant pressure, and a controlled build-up of pump pressure, While permanent internal monitoring and self-lubricating pumps ensure a long service life. The integrated 5.7 inch industrial touch screen displays critical operational information clearly and concisely for extra user-friendly operation.
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Wide working temperature ranges without the need to switch fluids
  • Highest performance with small footprint
  • Space-saving design optimises space utilization in your lab
  • 5.7 inch industrial color TFT touch screen
  • USB (host and device) and Ethernet connectivity
  • SD-Card slot
  • RS232 / optional RS485 / optional Profibus DP
  • Stand-by input
  • Easily removable venting grid
  • Available voltage versions: 3 x 400V/50Hz, 3 x 230V/50Hz, 3 x 230V/60Hz
Specifications
Specifications
Classification III (FL) according to DIN 12876-1
65 cm
-80°C to +250°C
PRESTO™ A80t
TFT Touchscreen
Ideal solution for highly precise, external temperature application
126 cm
250°C
43 cm
167 kg
Temperature Control System
3 x 230/400 V