missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ PrestoBlue™ Cell Viability Reagent
PrestoBlue™ Cell Viability Reagent
£205.00 - £483.00
Specifications
Cell Type | Bacteria, Eukaryotic Cells, Fungal Cells |
---|---|
Color | Blue |
Concentration | 10 X |
Content And Storage | Store at 4°C and protect from light. |
Description | PrestoBlue Cell Viability Reagent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12023725
|
Molecular Probes™
A13261 |
25 mL |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12083745
|
Molecular Probes™
A13262 |
100 mL |
£483.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Get high quality cell viability results and instant time savings with PrestoBlue™ Cell Viability Reagent. PrestoBlue™ is a ready to use cell permeable resazurin-based solution that functions as a cell viability indicator by using the reducing power of living cells to quantitatively measure the proliferation of cells. When added to cells, the PrestoBlue™ reagent is modified by the reducing environment of the viable cell and turns red in color, becoming highly fluorescent. This color change can be detected using fluorescence or absorbance measurements.The PrestoBlue™ Cell Viability Reagent allows you to:
10-minute Incubations
The PrestoBlue™ Cell Viability Reagent is the only live-cell viability reagent on the market able to provide robust data typically in as little as 10 minutes following reagent addition. All other resazurin-based cell viability reagents on the market require a one to four hour incubation with the reagent prior to obtaining data.
High Quality Results
Based on the cell type and incubation time, PrestoBlue™ Cell Viability Reagent can detect as few as 12 cells per well in a 384 well plate. After ten minutes of incubation the PrestoBlue™ Reagent can linearly detect cells down to 98 cells⁄well with a Z’ value of greater than 0.5.
Homogenous Assay Format
The PrestoBlue™ Cell Viability Reagent is an addition-only reagent. Without the cell lysis or solubilization steps required by other assays on the market, hands-on time is minimized.
Live Cell Assay
The PrestoBlue™ Cell Viability Reagent does not require cell lysis. Cells may be incubated for up to 24 hours in the presence of PrestoBlue™ Cell Viability Product. Simply remove the reagent from the cells and replace it with growth medium for further proliferation.
Specifications
Bacteria, Eukaryotic Cells, Fungal Cells | |
10 X | |
PrestoBlue Cell Viability Reagent | |
Viability Assay | |
Reagent | |
590-615 nm | |
Liquid | |
10 min. to 2 hr. | |
High-throughput Compatible |
Blue | |
Store at 4°C and protect from light. | |
Colorimetric, Fluorescence | |
Microplate Reader, Spectrophotometer, HTS Reader | |
Resazurin | |
535-560 nm | |
96-well plate, Cuvettes, 384-well plate | |
Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.