Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Brand: Thermo Scientific™ D8830
Code : A0
Additional Details : Weight : 0.80000kg
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.Specifications
Barnstead Pre-Filter MQ Cartridge, 0.5 Micron | |
Milli-Q Millipore Water Systems | |
Pre-Filter MQ Cartridge |