Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Brand:  Thermo Scientific™ D63111
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15322178
  • £148.86 / 1 unit
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
Specifications
Cellulose Acetate Prefilter, 5 Micron, for RoPure Reverse Osmosis Water Systems
RoPure Reverse Osmosis Water Systems
Cellulose Acetate