missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine, 98%, Thermo Scientific Chemicals

Catalog Number p-7077607
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
721-50-6
C13H20N2O
220.32
MFCD00048681
MVFGUOIZUNYYSO-UHFFFAOYNA-N
prilocaine, propitocaine, citanest, prilocainum, prilocaina, astra 1515, n-2-methylphenyl-2-propylamino propanamide, o-methyl-2-propylaminopropionanilide, propanamide, n-2-methylphenyl-2-propylamino, astra 1512
4906
CHEBI:8404
N-(2-methylphenyl)-2-(propylamino)propanamide
CCCNC(C)C(=O)NC1=CC=CC=C1C
This item is not returnable. View return policy
721-50-6
C13H20N2O
220.32
MFCD00048681
MVFGUOIZUNYYSO-UHFFFAOYNA-N
prilocaine, propitocaine, citanest, prilocainum, prilocaina, astra 1515, n-2-methylphenyl-2-propylamino propanamide, o-methyl-2-propylaminopropionanilide, propanamide, n-2-methylphenyl-2-propylamino, astra 1512
4906
CHEBI:8404
N-(2-methylphenyl)-2-(propylamino)propanamide
CCCNC(C)C(=O)NC1=CC=CC=C1C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 36.0°C to 38.7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 220.31
Percent Purity 98%
Chemical Name or Material Prilocaine
compliance-icons
Product Identifier
  • Prilocaine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'