missing translation for 'onlineSavingsMsg'
Learn More

Primary Drinking Water Metals For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep EP-8-500

Additional Details : CAS Number : 7440-39-3 Weight : 0.50000kg Transport : UN number : 3264 Chem class : 8 Pack group : III


 View more versions of this product

Product Code. 15628295

  • £310.00 / 500mL
Estimated Shipment: 29-08-2024
to see stock.

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba100μg/mL
Ag10μg/mL
As10μg/mL
Cr10μg/mL
Pb10μg/mL
Cd5μg/mL
Se5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Primary Drinking Water Metals
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
2% HNO3
Drinking Water Pollutant Standard
500 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Various
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Primary Drinking Water Metals

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only