missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Mouse Hippocampal Neurons

Primary Mouse Hippocampal Neurons

Brand:  Gibco™ A15587

731.43 GBP valid until 2025-01-17
Use promo code "21645" to get your promotional price.


Product Code. 15363816

  • £694.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco™ Primary Mouse Hippocampal Neurons are isolated and cryopreserved from C57BL/6 embryonic day-17 mice. A flexible, ready-to-use, and quality alternative to fresh neurons, each vial contains 1 million cells per mL. These neurons consistently yield post-thaw viability in the range of 50-80%, have high neuronal purity, and are validated for functionality.

  • High viability: post-thaw viability of 50% or greater
  • High purity: greater than 98% neuronal phenotype
  • Functionally validated: tested for long-term survival, neurotransmitter response, and transduction
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store in liquid nitrogen, vapor-phase.
    Neural Cells
    Cryopreserved
    Hippocampus
    Mouse Hippocampal Neuron
    Mouse strain is C57BL/6
    Fetal
    Mammalian Cell Culture
    Mouse
    1 Million
    1 x 106 cells
    Sterility, Viability, Mycoplasma
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.