Learn More
Invitrogen™ PRIMER 10 UMOL
Custom Primer
Brand: Invitrogen™ 10629087
11.07 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
Primer, 10μmol | |
Primer | |
10 μmol |