Learn More
Invitrogen™ PRIMER 5' TAM
Custom Primer
Brand: Invitrogen™ 11773025
130.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' TAM Primer, 1μmol | |
5' TAM Primer | |
5' Labeled | |
1 μmol |