Learn More
Invitrogen™ PRIMER URACIL 25 NMOL
Custom Primer
Brand: Invitrogen™ 10623064
6.95 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
Uracil Primer, 25 nmol | |
Uracil Primer | |
25 nmol |