Learn More
Thermo Scientific™ PrimeStore Molecular Transport Medium
Non-hazardous, inactivation media for viral collection and transport with Inactivation media for nucleic acid testing and molecular diagnostics.
£286.00
Specifications
Closure Color | Clear |
---|---|
Compatibility | The PS-MTM system is a collection, preservation, transport, and storage system for influenza RNA and MTB DNA. Extraction and purification of nucleic acids have been validated on several manual spin column kits (PrimeXtract™, RNAqueous Micro Kit, Viral RNA Mini Kit, QiaAMP DNA Mini Kit) and automated magnetic bead extraction kits (NucliSENS EasyMAG and MagNA Pure 96 System using the DNA Bacterial/Viral small volume kit). The user is responsible for validating additional extraction and purification kits and platforms. |
For Use With (Application) | Intended for stabilization, transportation, and inactivation of infectious unprocessed nasal washes, nasopharyngeal, oral/throat swabs suspected of containing influenza A virus RNA. PS-MTM is also intended for the stabilization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples. |
Quantity | 50/Pk. |
Storage Requirements | Room temperature, 2°C to 25°C |
Product Code | Brand | Description | Fill Volume | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Fill Volume | Volume (Metric) | Price | Quantity & Availability | |||
17808377
|
Thermo Scientific™
R13905 |
Primestore MTM Collection Tube (1.5 mL) | 1.5 mL | 5 mL |
£286.00
Pack of 200 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Stabilize, inactivate and transport infectious unprocessed nasal washes, nasopharyngeal, oral / throat swabs suspected of containing influenza A virus RNA with Longhorn PrimeStore Molecular Transport Medium™ (PS-MTM).
PrimeStore Molecular Transport Medium™ is also intended for the stabilization, transportation, and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.
Specimen collection and transport is a key component in the molecular detection of influenza A and MTB. PS-MTM is a self-contained, ‘ready-to-use’ system that allows for the stabilization and safe transport of clinical samples at ambient temperature from the collection site to the laboratory.
Specifications
Clear | |
Intended for stabilization, transportation, and inactivation of infectious unprocessed nasal washes, nasopharyngeal, oral/throat swabs suspected of containing influenza A virus RNA. PS-MTM is also intended for the stabilization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples. | |
Room temperature, 2°C to 25°C |
The PS-MTM system is a collection, preservation, transport, and storage system for influenza RNA and MTB DNA. Extraction and purification of nucleic acids have been validated on several manual spin column kits (PrimeXtract™, RNAqueous Micro Kit, Viral RNA Mini Kit, QiaAMP DNA Mini Kit) and automated magnetic bead extraction kits (NucliSENS EasyMAG and MagNA Pure 96 System using the DNA Bacterial/Viral small volume kit). The user is responsible for validating additional extraction and purification kits and platforms. | |
50/Pk. |
Distributed globally by Thermo Fisher Scientific