Learn More
Zeiss™ Primovert Inverted Microscope with Phototube
Compact inverted routine microscope that combines efficiency and optical performance. This inverted microscope is configured for transmitted light brightfield and phase contrast. This system is ideal for cell culture, routine work and quick & easy imaging.
£3230.00 - £6425.00
Specifications
For Use With (Application) Cell Culture, Routine Work and Quick and Easy Imaging
View More Specs
Products 2
Product Code Brand Objective Types Price Quantity & Availability  
Product Code Brand Objective Types Price Quantity & Availability  
10636717
View Documents
Zeiss™
491206-0002-000
4X and 10X
£3230.00
Each
N/A
Currently Unavailable: We have no information when this product will be available
View alternative products
15168229
View Documents
Zeiss™
491206-0011-000
4X, 10X, 20X and 40X
£6425.00
Each
N/A
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • The Primovert is perfectly suited to your cell culture laboratory.
  • It enables fast, efficient investigations of both unstained cells in phase contrast and GFP-labeled cells in fluorescence contrast.
  • It fits straight into your laminar flow cabinet to work directly in a sterile environment.
  • The Primovert is easy to use and with little set up the system is ready to use out of the box.
  • Automatic light source switch-off (in addition to manual on/off-function): Primo Vert switches off the light automatically after 15 min - saving energy and protecting the stand
  • Modular illumination: halogen or long lasting LED
  • High quality material: the stand is made of metal and all operating elements are coated with a special touch-friendly rubber compound
  • Universal phase slider (optional) for all objectives: for the ease of use there is one phase ring (Ph1) available for 10x, 20x and 40x - this means not having to change the position of the phase slider when changing objectives. In short: time-saving and economical
  • Increased working distance thanks to easy removal of the condenser, e.g., for roller-bottles
  • Objective indicators for fast identification of magnification
  • Integrated carrying handle
  • Award-winning design The new Primo Vert inverted microscope is focused on the essentials: excellent optics, high-durability quality materials, ease of use and an appealing design.
  • The Primo Vert addresses a wide range of applications: from routine laboratories for live cell inspection to cutting edge research laboratories which require - in addition to research microscopes - compact and reliable routine microscopes for quick and efficient viewing of living cells.
  • Typical applications include cancer and HIV research, human, animal and plant genetics, and cell biology in general.
Specifications
Specifications
Cell Culture, Routine Work and Quick and Easy Imaging