Learn More
Epredia™ PrintMate™ AS Cassette Printer Accessories
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Brand:  Epredia™ B81300033
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12963815
  • £5155.00 / Each
Estimated Shipment: 08-08-2024
to see stock.
Description
Description
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
  • Specifications
    Specifications
    Computer
    15 in. All-in-one PC
    PrintMate™ AS Cassette Printer, SlideMate™ Slide Printer