Learn More
Epredia™ PrintMate™ AS Cassette Printer
The small footprint of the Epredia™ SlideMateTM AS Slide Printer brings the convenience of on-demand slide printing directly to the microtome. The SlideMate AS Slide Printer drastically reduces the chance for patient identification errors by allowing the laboratory to print accurate information at the moment the slide is required.
£29170.00 - £69000.00
Specifications
Electrical Requirements 100 to 240 VAC, 50/60 Hz, 150 VA (512 BTU/h)
View More Specs
Products 6
Product Code Brand Description Includes Price Quantity & Availability  
Product Code Brand Description Includes Price Quantity & Availability  
15488891
View Documents
Epredia™
A83800001
PrintMate AS 150 Cassette Printer PrintMate AS
USB cable
Power cable
2 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
£29170.00
Each
In Stock
to see stock.
Add to basket
15498891
View Documents
Epredia™
A83800002
PrintMate AS 450 Cassette Printer PrintMate AS
USB cable
Power cable
6 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
£39700.00
Each
In Stock
to see stock.
Add to basket
15408901
View Documents
Epredia™
A83800003
PrintMate AS 900 Cassette Printer PrintMate AS
USB cable
Power cable
12 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
£63520.00
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
15418901
View Documents
Epredia™
A83800004
PrintMate AS 150 Cassette Printing System PrintMate AS
USB cable
Power cable
2 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
All-in-one PC
Digital imager
£30965.00
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
15441021
View Documents
Epredia™
A83800005
PrintMate AS 450 Cassette Printing System PrintMate AS
USB cable
Power cable
6 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
All-in-one PC
Digital imager
£42455.00
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
15448901
View Documents
Epredia™
A83800006
PrintMate AS 900 Cassette Printing System PrintMate AS
USB cable
Power cable
12 cassette hoppers
One roll of tape (installed)
Epredia™ LabWriter™ software CD
Operator guide
All-in-one PC
Digital imager
£69000.00
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
Highlights:
  • Prints each cassette in 10 seconds or less
  • Small footprint allows the printer to fit easily next to the grossing station
  • Print only the cassettes you need, right at the point of use
  • Print barcodes on cassettes to automate downstream processes
  • Eliminate manual handwriting processes
Specifications
Specifications
100 to 240 VAC, 50/60 Hz, 150 VA (512 BTU/h)