missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pristane, MP Biomedicals™
Click to view available options
Quantity:
5 mL
Description
Pristane is known to induce autoimmune diseases in rodents. It is used in research to understand the pathogenesis of rheumatoid arthritis and lupus, as a non-antigenic arthritogenic adjuvant and plasmacytomas.
Specifications
CAS | 1921-70-6 |
Density | 0.783g/mL at 20°C (Lit.) |
Boiling Point | 68°C (0.001 mmHg (literature)) |
Molecular Formula | C19H40 |
Refractive Index | n20/D 1.438 (Lit.) |
Quantity | 5 mL |
Synonym | pristane, norphytane, bute hydrocarbon, norphytan, pristan, pentadecane, 2,6,10,14-tetramethyl, meso-pristane, 2,6,10,10-tetramethylpentadecane, meso-form, norphytane, robuoy |
Solubility Information | Soluble in ether, petroleum ether, benzene, chloroform, carbon tetrachloride. |
InChI Key | XOJVVFBFDXDTEG-UHFFFAOYSA-N |
SMILES | CC(C)CCCC(C)CCCC(C)CCCC(C)C |
Show More |
Safety and Handling
Product Identifier
- Pristane
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 2,6,10,14-tetramethyl-Pentadecane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'